Merck
  • Cardiac responses to hypercapnia in larval zebrafish (Danio rerio): the links between CO2 chemoreception, catecholamines and carbonic anhydrase.
Cardiac responses to hypercapnia in larval zebrafish (Danio rerio): the links between CO2 chemoreception, catecholamines and carbonic anhydrase.
The Journal of experimental biology (2014-07-27)
Scott Miller, Jacob Pollack, Julia Bradshaw, Yusuke Kumai, Steve F Perry
ABSTRACT
The ontogeny of carbon dioxide (CO2) sensing in zebrafish (Danio rerio) has not been examined. In this study, CO2-mediated increases in heart rate were used to gauge the capacity of zebrafish larvae to sense CO2. CO2 is thought to be detected via neuroepithelial cells (NECs), which are homologous to mammalian carotid body glomus cells. Larvae at 5 days post-fertilization (d.p.f.) exhibited tachycardia when exposed for 30 min to 0.75% CO2 (~5.63 mmHg); at 7 d.p.f., tachycardia was elicited by 0.5% CO2 (~3.75 mmHg). Based on pharmacological evidence using β-adrenergic receptor (β-AR) antagonists, and confirmed by β1-AR translational gene knockdown using morpholinos, the reflex tachycardia accompanying hypercapnia was probably mediated by the interaction of catecholamines with cardiac β1 receptors. Because the cardiac response to hypercapnia was abolished by the ganglionic blocker hexamethonium, it is probable that the reflex cardio-acceleration was mediated by catecholamines derived from sympathetic adrenergic neurons. Owing to its likely role in facilitating intracellular acidification during exposure to hypercapnia, it was hypothesized that carbonic anhydrase (CA) is involved in CO2 sensing, and that inhibition of CA activity would blunt the downstream responses. Indeed, the cardiac response to hypercapnia (0.75% CO2) was reduced in fish at 5 d.p.f. exposed to acetazolamide, a CA inhibitor, and in fish experiencing zCAc (CA2-like a) knockdown. Successful knockdown of zCAc was confirmed by CA activity measurements, western blotting and immunocytochemistry. Co-injection of embryos with zCAc morpholino and mRNA modified at the morpholino binding site restored normal levels of CA activity and protein levels, and restored (rescued) the usual cardiac responses to hypercapnia. These data, combined with the finding that zCAc is expressed in NECs located on the skin, suggest that the afferent limb of the CO2-induced cardiac reflex in zebrafish larvae is initiated by coetaneous CO2-sensing neuroepithelial cells.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M