Merck
  • Stability study of full-length antibody (anti-TNF alpha) loaded PLGA microspheres.
Stability study of full-length antibody (anti-TNF alpha) loaded PLGA microspheres.
International journal of pharmaceutics (2014-05-06)
S Marquette, C Peerboom, A Yates, L Denis, I Langer, K Amighi, J Goole
ABSTRACT
Antibodies (Abs) require the development of stable formulations and specific delivery strategies given their susceptibility to a variety of physical and chemical degradation pathways. In this study, the encapsulation of an antibody into polylactide-co-glycolide (PLGA) based microspheres was explored to obtain a controlled-release of the incorporated drug. In order to avoid stability issues, a solid-in-oil-in-water (s/o/w) method was preferred. The solid phase was made of anti-TNF alpha monoclonal antibody (MAb) spray-dried microparticles, and the PLGA microspheres were produced using two different polymers (i.e., Resomer(®) RG505 and Resomer(®) RG755S). The stability of the MAb incorporated into the microspheres was investigated under three conditions (5 ± 3°C, 25 ± 2°C/60% RH and 40 ± 2°C/75% RH) for 12 weeks. During this stability study, it was demonstrated that the MAb loaded PLGA microspheres were stable when stored at 5 ± 3°C and that the Resomer(®) RG755S, composed of 75%(w/w) lactic acid as PLGA, was preferred to preserve the stability of the system. Storage at temperatures higher than 5°C led to antibody stability issues such as aggregation, fragmentation and loss of activity. The release profiles were also altered. Physical ageing of the system associated with changes in the glass transition temperature and enthalpy of relaxation was noticed during the storage of the MAb loaded PLGA microspheres.
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Supelco
Dichloromethane, analytical standard
Histidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material