Merck
  • A new approach to standardize multicenter studies: mobile lab technology for the German Environmental Specimen Bank.
A new approach to standardize multicenter studies: mobile lab technology for the German Environmental Specimen Bank.
PloS one (2014-08-21)
Dominik Lermen, Daniel Schmitt, Martina Bartel-Steinbach, Christa Schröter-Kermani, Marike Kolossa-Gehring, Hagen von Briesen, Heiko Zimmermann
ABSTRACT
Technical progress has simplified tasks in lab diagnosis and improved quality of test results. Errors occurring during the pre-analytical phase have more negative impact on the quality of test results than errors encountered during the total analytical process. Different infrastructures of sampling sites can highly influence the quality of samples and therewith of analytical results. Annually the German Environmental Specimen Bank (ESB) collects, characterizes, and stores blood, plasma, and urine samples of 120-150 volunteers each on four different sampling sites in Germany. Overarching goal is to investigate the exposure to environmental pollutants of non-occupational exposed young adults combining human biomonitoring with questionnaire data. We investigated the requirements of the study and the possibility to realize a highly standardized sampling procedure on a mobile platform in order to increase the required quality of the pre-analytical phase. The results lead to the development of a mobile epidemiologic laboratory (epiLab) in the project "Labor der Zukunft" (future's lab technology). This laboratory includes a 14.7 m(2) reception area to record medical history and exposure-relevant behavior, a 21.1 m(2) examination room to record dental fillings and for blood withdrawal, a 15.5 m(2) biological safety level 2 laboratory to process and analyze samples on site including a 2.8 m(2) personnel lock and a 3.6 m2 cryofacility to immediately freeze samples. Frozen samples can be transferred to their final destination within the vehicle without breaking the cold chain. To our knowledge, we herewith describe for the first time the implementation of a biological safety laboratory (BSL) 2 lab and an epidemiologic unit on a single mobile platform. Since 2013 we have been collecting up to 15.000 individual human samples annually under highly standardized conditions using the mobile laboratory. Characterized and free of alterations they are kept ready for retrospective analyses in their final archive, the German ESB.
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%