Merck
  • Ca2+ influx and ATP release mediated by mechanical stretch in human lung fibroblasts.
Ca2+ influx and ATP release mediated by mechanical stretch in human lung fibroblasts.
Biochemical and biophysical research communications (2014-09-27)
Naohiko Murata, Satoru Ito, Kishio Furuya, Norihiro Takahara, Keiji Naruse, Hiromichi Aso, Masashi Kondo, Masahiro Sokabe, Yoshinori Hasegawa
ABSTRACT
One cause of progressive pulmonary fibrosis is dysregulated wound healing after lung inflammation or damage in patients with idiopathic pulmonary fibrosis and severe acute respiratory distress syndrome. The mechanical forces are considered to regulate pulmonary fibrosis via activation of lung fibroblasts. In this study, the effects of mechanical stretch on the intracellular Ca(2+) concentration ([Ca(2+)]i) and ATP release were investigated in primary human lung fibroblasts. Uniaxial stretch (10-30% in strain) was applied to fibroblasts cultured in a silicone chamber coated with type I collagen using a stretching apparatus. Following stretching and subsequent unloading, [Ca(2+)]i transiently increased in a strain-dependent manner. Hypotonic stress, which causes plasma membrane stretching, also transiently increased the [Ca(2+)]i. The stretch-induced [Ca(2+)]i elevation was attenuated in Ca(2+)-free solution. In contrast, the increase of [Ca(2+)]i by a 20% stretch was not inhibited by the inhibitor of stretch-activated channels GsMTx-4, Gd(3+), ruthenium red, or cytochalasin D. Cyclic stretching induced significant ATP releases from fibroblasts. However, the stretch-induced [Ca(2+)]i elevation was not inhibited by ATP diphosphohydrolase apyrase or a purinergic receptor antagonist suramin. Taken together, mechanical stretch induces Ca(2+) influx independently of conventional stretch-sensitive ion channels, the actin cytoskeleton, and released ATP.
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Ruthenium Red, Technical grade
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO