Merck
  • Maternal perinatal undernutrition modifies lactose and serotranferrin in milk: relevance to the programming of metabolic diseases?
Maternal perinatal undernutrition modifies lactose and serotranferrin in milk: relevance to the programming of metabolic diseases?
American journal of physiology. Endocrinology and metabolism (2015-01-01)
J S Wattez, A Delmont, M Bouvet, O Beseme, S Goers, F Delahaye, C Laborie, J Lesage, B Foligné, C Breton, C C Metges, D Vieau, F Pinet
ABSTRACT
A close link between intrauterine growth restriction and development of chronic adult diseases such as obesity, diabetes, and hypertension has been established both in humans and animals. Modification of growth velocity during the early postnatal period (i.e., lactation) may also sensitize to the development of metabolic syndrome in adulthood. This suggests that milk composition may have long-lasting programming/deprogramming metabolic effects in the offspring. We therefore assess the effects of maternal perinatal denutrition on breast milk composition in a food-restricted 50% (FR50) rat model. Monosaccharides and fatty acids were characterized by gas chromatography, and proteins were profiled by surface-enhanced laser desorption/ionization-time-of-flight analysis in milk samples from FR50 and control rat dams. Milk analysis of FR50 rats demonstrated that maternal undernutrition decreases lactose concentration and modulates lipid profile at postnatal day 10 by increasing the unsaturated fatty acids/saturated fatty acids and diminishes serotransferrin levels at postnatal day 21. Our data indicate that maternal perinatal undernutrition modifies milk composition both quantitatively and qualitatively. These modifications by maternal nutrition open new perspectives to identify molecules that could be used in artificial milk to protect from the subsequent development of metabolic diseases.
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder