Merck
  • Ribosomic DNA intergenic spacer 1 region is useful when identifying Candida parapsilosis spp. complex based on high-resolution melting analysis.
Ribosomic DNA intergenic spacer 1 region is useful when identifying Candida parapsilosis spp. complex based on high-resolution melting analysis.
Medical mycology (2014-05-23)
Sara Gago, Ana Alastruey-Izquierdo, Marco Marconi, María José Buitrago, Arnaud Kerhornou, Paul J Kersey, Emilia Mellado, Manuel Cuenca-Estrella, Juan Luis Rodríguez-Tudela, Isabel Cuesta
ABSTRACT
The epidemiology of Candida parapsilosis and the closely related species C. orthopsilosis and C. metapsilosis has changed in recent years, justify the need to identify this complex at the species level. In this study we investigate the intergenic spacer 1 (IGS1) of the ribosomal DNA (rDNA) to evaluate the utility of this gene region as a phylogenetic molecular marker and the suitability of a high-resolution melting (HRM) strategy based on this region for identification of members of the C. parapsilosis spp. complex. We sequenced the IGS1 and the internal transcribed spacer (ITS) regions of the rDNA from 33 C. parapsilosis sensu lato strains. Although both regions are useful in identifying species, comparative sequence analysis showed that the diversity in the IGS1 region was higher than in the ITS sequences. We also developed an HRM analysis that reliably identifies C. parapsilosis spp. complex based on the amplification of 70 bp in the IGS1 region. All isolates were correctly identified with a confidence interval >98%. Our results demonstrate that HRM analysis based on the IGS1 region is a powerful tool for distinguishing C. parapsilosis from cryptic species.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, puriss. p.a., ACS reagent, ≥99.8% (chloroform + ethanol, GC)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer