Merck
  • Skin dryness in apparently healthy human skin is associated with decreased expression of bleomycin hydrolase in the stratum corneum.
Skin dryness in apparently healthy human skin is associated with decreased expression of bleomycin hydrolase in the stratum corneum.
Clinical and experimental dermatology (2014-12-17)
E D Son, Y Kim, K M Joo, H J Kim, E Lee, G W Nam, E G Cho, M Noh, J H Chung, S Y Byun, T R Lee
ABSTRACT
Maintenance of water balance in the stratum corneum (SC) is determined by the content of intercellular lipids and natural moisturizing factors (NMFs) in corneocytes. To investigate the association between the NMFs and (pro)filaggrin and the proteases responsible for the processing of (pro)filaggrin to NMFs in the SC of hydrated and dry skin areas of healthy human subjects. The SC hydration state and the transepidermal water loss (TEWL) were measured using a Corneometer and a Tewameter, respectively. Proteases, (pro)filaggrin and NMFs were extracted from SC samples obtained by tape-stripping of the tested skin. Expression levels of (pro)filaggrin were determined by dot blotting and western blotting, and total NMFs by ultra-high performance liquid chromatography. Expression of the proteases caspase-14, calpain-1 and bleomycin hydrolase was measured by western blotting. The levels of (pro)filaggrin were not significantly different between hydrated and dry skin, whereas the level of total NMFs was significantly reduced in dry skin. A negative correlation between (pro)filaggrin and NMFs was found in dry skin (Pearson correlation coefficient r = - 0.57, *P < 0.05). Bleomycin hydrolase expression was significantly decreased in the SC of dry skin. These results suggest that the low hydration state of dry skin may be due to the reduction in (pro)filaggrin degradation caused by decreased bleomycin hydrolase expression.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized