Merck
  • Plasma protein binding, pharmacokinetics, tissue distribution and CYP450 biotransformation studies of fidarestat by ultra high performance liquid chromatography-high resolution mass spectrometry.
Plasma protein binding, pharmacokinetics, tissue distribution and CYP450 biotransformation studies of fidarestat by ultra high performance liquid chromatography-high resolution mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Roshan M Borkar, Murali Mohan Bhandi, Ajay P Dubey, Prajwal P Nandekar, Abhay T Sangamwar, Sanjay K Banerjee, R Srinivas
ABSTRACT
Fidarestat, an aldose reductase inhibitor, has been used for the treatment of the diabetic associated complications such as retinopathy, neuropathy and nephropathy. To better understand the metabolism and pharmacokinetics of fidarestat, we have evaluated plasma protein binding, pharmacokinetics, tissue distribution of the drug and its conjugated metabolites and CYP450 biotransformation by liquid chromatography-high resolution mass spectrometry. Effective chromatographic separation of fidarestat and hydrochlorothiazide (IS) in rat plasma and tissues was achieved on Hypersil gold C-18 column in an isocratic elution mode. For detection, a high-resolution Orbitrap mass spectrometer with heated electrospray ionization inlet in the negative ion mode was used. High-resolution extracted ion chromatograms for each analyte were obtained by processing the full-scan MS mode with 5 ppm mass tolerance. The impact of plasma protein binding with the drug and conjugated metabolites of the drug on pharmacokinetics has been determined. The study indicated that 9.5% of free form of fidarestat may be pharmacologically active and the Cmax for free fidarestat was found to be 80.30 ± 6.78 ng/mL. The AUC0-t and AUC0-∞ were found to be 185.46 ± 32 and 195.92 ± 15.06 ng h/mL, respectively. Among tissues, the maximum observed distribution was found to be in kidney followed by liver and heart. Docking experiments and in vitro CYP450 reaction phenotyping revealed that two CYP1A2 and CYP2D6 are involved in the phase I metabolism of fidarestat. Oxidative deamination and N/O glucuronidation are the major phase I and phase II metabolites, respectively. In vitro CYP450 inhibition assay of fidarestat for drug-drug interaction showed weak inhibition and may not alter pharmacokinetics, distribution or clearance of other co-administered drug.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Adenosine 3′-monophosphate, from yeast
Supelco
Warfarin, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC