Merck
Synthetic isoxazole as antiplatelet agent.
Platelets (2013-07-12)
Margarita Gutiérrez, Jessica Amigo, Eduardo Fuentes, Ivan Palomo, Luis Astudillo
ABSTRACT
Abstract Nine synthetic isoxazoles were evaluated as antiplatelet agents and studied the possible mechanism of more active compound. The initial screening was evaluating all compounds against platelet aggregation assays. The most active compound was isoxazole 8 showing an inhibition of platelet aggregation around 70%. In subsequent experiments, ADP and collagen were used as agonists to explore the possible inhibitory mechanisms of isoxazole 8 in platelet aggregation and secretion. We reported the effect of isoxazole 8 for reducing the expression of inflammatory markers, such as soluble CD40 ligand (sCD40L) and soluble P-selectin (sP-selectin), on activated platelets. Of this form, an inhibition of sCD40L and sP-selectin can prevent the onset of an atherosclerotic lesion.
Product Description
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Adenosine 5′-diphosphate, ≥95% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard