Merck
  • The tumor necrosis factor superfamily members TWEAK, TNFSF15 and fibroblast growth factor-inducible protein 14 are upregulated in proliferative diabetic retinopathy.
The tumor necrosis factor superfamily members TWEAK, TNFSF15 and fibroblast growth factor-inducible protein 14 are upregulated in proliferative diabetic retinopathy.
Ophthalmic research (2015-02-14)
Ahmed M Abu El-Asrar, Gert De Hertogh, Mohd Imtiaz Nawaz, Mohammad Mairaj Siddiquei, Kathleen Van den Eynde, Ghulam Mohammad, Ghislain Opdenakker, Karel Geboes
ABSTRACT
Tumor necrosis factor-like weak inducer of apoptosis (TWEAK) and tumor necrosis factor superfamily member 15 (TNFSF15), members of the TNF superfamily, play important roles in the modulation of inflammation and neovascularization. TWEAK activity is mediated via binding to fibroblast growth factor-inducible molecule 14 (Fn14). We investigated the expression of TWEAK, Fn14 and TNFSF15 and the correlation between TWEAK levels and the levels of the inflammatory biomarker soluble intercellular adhesion molecule-1 (sICAM-1) in proliferative diabetic retinopathy (PDR). In addition, we examined the expression of FN14 and TNFSF15 in retinas of diabetic rats. Vitreous samples from 34 PDR and 23 nondiabetic patients were studied by enzyme-linked immunosorbent assay and Western blot analysis. Epiretinal membranes from 14 patients with PDR were studied by immunohistochemistry. The retinas of rats were examined by Western blot analysis. We identified a significant increase in the expression of TWEAK, Fn14, TNFSF15 and sICAM-1 in vitreous samples from PDR patients compared to controls. A significant positive correlation was found between levels of TWEAK and levels of sICAM-1 (r = 0.3, p = 0.02). In epiretinal membranes, TWEAK and TNFSF15 protein expression was confined to vascular endothelial cells, monocytes/macrophages and myofibroblasts. Significant positive correlations were observed between the number of blood vessels expressing CD34 and the number of blood vessels expressing TWEAK (r = 0.670; p = 0.017) and TNFSF15 (r = 0.784; p = 0.001). The expression level of TNFSF15 was upregulated in the retinas of diabetic rats, whereas Fn14 was not upregulated. Our findings suggest that TNFSF15 and the TWEAK/Fn14 pathway are novel mediators involved in persistent inflammation and modulation of pathological neovascularization associated with PDR.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)