Merck
  • Bio-inspired solid phase extraction sorbent material for cocaine: a cross reactivity study.
Bio-inspired solid phase extraction sorbent material for cocaine: a cross reactivity study.
Talanta (2014-08-28)
Camilla Montesano, Manuel Sergi, German Perez, Roberta Curini, Dario Compagnone, Marcello Mascini
ABSTRACT
The binding specificity of a bio-inspired hexapeptide (QHWWDW) versus cocaine and four other drugs such as 3,4-methylenedioxy-N-methylamphetamine (MDMA), 3,4-methylenedioxy-N-ethylamphetamine (MDEA), phencyclidine and morphine was computationally studied and then experimentally confirmed in solid phase extraction (SPE) followed by liquid chromatography-mass spectrometry (LC/MS) detection. In simulation, the hexapeptide-drug complexes were docked with different scoring functions and considering pH chemical environment. In experimental, the cross reactivity of the selected hexapeptide was tested as SPE sorbent versus cocaine and other four drugs using buffer solutions at pH 4 and 7. Significant differences in specific retention were found between cocaine (97% of recovery) and both morphine (45% of recovery) and phencyclidine (60% of recovery), but less for ecstasies (average recovery 69%). In agreement with docking simulation, the hexapeptide showed the highest recovery with best specificity versus cocaine at pH 7 with an experimentally binding constant of 2.9 × 10(6)M(-1). The bio-inspired sorbent material analytical performances were compared with a commercial reversed phase cartridge confirming the hexapeptide specificity to cocaine and validating simulated data.
Sigma-Aldrich
N-Methyldiethanolamine, ≥99%
Sigma-Aldrich
Cocaine free base