Merck
  • Nod factor receptors form heteromeric complexes and are essential for intracellular infection in medicago nodules.
Nod factor receptors form heteromeric complexes and are essential for intracellular infection in medicago nodules.
The Plant cell (2014-10-30)
Sjef Moling, Anna Pietraszewska-Bogiel, Marten Postma, Elena Fedorova, Mark A Hink, Erik Limpens, Theodorus W J Gadella, Ton Bisseling
ABSTRACT
Rhizobial Nod factors are the key signaling molecules in the legume-rhizobium nodule symbiosis. In this study, the role of the Nod factor receptors NOD FACTOR PERCEPTION (NFP) and LYSIN MOTIF RECEPTOR-LIKE KINASE3 (LYK3) in establishing the symbiotic interface in root nodules was investigated. It was found that inside Medicago truncatula nodules, NFP and LYK3 localize at the cell periphery in a narrow zone of about two cell layers at the nodule apex. This restricted accumulation is narrower than the region of promoter activity/mRNA accumulation and might serve to prevent the induction of defense-like responses and/or to restrict the rhizobium release to precise cell layers. The distal cell layer where the receptors accumulate at the cell periphery is part of the meristem, and the proximal layer is part of the infection zone. In these layers, the receptors can most likely perceive the bacterial Nod factors to regulate the formation of symbiotic interface. Furthermore, our Förster resonance energy transfer-fluorescence lifetime imaging microscopy analysis indicates that NFP and LYK3 form heteromeric complexes at the cell periphery in M. truncatula nodules.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%