Merck
  • Development of a CZE method for the quantification of pseudoephedrine and cetirizine.
Development of a CZE method for the quantification of pseudoephedrine and cetirizine.
Journal of chromatographic science (2013-09-24)
Ahmed O Alnajjar, Abubakr M Idris
ABSTRACT
Pseudoephedrine and cetirizine have been combined in dosage forms with more therapeutic benefits when compared with single-drug treatment. The current manuscript reports the development of the first capillary zone electrophoresis (CZE) assay method for that combination. The effects of pH and buffer concentration on resolution, noise, migration time and peak area were examined employing experimental design approach. The analytes were electropherographed into a 50.2 cm-long and 50 µm i.d. fused-silica capillary column using 10 mmol/L borate at pH 8.3 with a potential of 25 kV at 25°C and UV detection at 214 nm. The method was successfully validated in order to verify its suitability for pharmaceutical analysis for the purposes of quality control. Over previous high-performance liquid chromatographic methods, the current CZE method features the benefits of the use of cost-effective electrolyte, besides high sample throughput (11 samples/h). Furthermore, other analytical results including linear dynamic ranges, recovery (96.9-98.1%), intra- and interday precision (relative standard deviation ≤ 1.70%) as well as the limits of detection and quantification (≤2.65 µg/mL) were all satisfactory for the intended purpose.
Product Description
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%