Merck
  • Conformational analysis of morpholine studied using Raman spectroscopy and density functional theoretical calculations.
Conformational analysis of morpholine studied using Raman spectroscopy and density functional theoretical calculations.
Chemical Physics Letters (2015)
SenGupta S, et al.