Merck
  • Oncogenic miR-23a in Pancreatic Ductal Adenocarcinogenesis Via Inhibiting APAF1.
Oncogenic miR-23a in Pancreatic Ductal Adenocarcinogenesis Via Inhibiting APAF1.
Digestive diseases and sciences (2015-02-24)
Ning Liu, Yuan-Yuan Sun, Xiao-Wen Zhang, Sheng Chen, Ye Wang, Zhao-Xiong Zhang, Shao-Wei Song, Guang-Bin Qiu, Wei-Neng Fu
ABSTRACT
miR-23a, which participates in invasion of pancreatic ductal adenocarcinoma cells into the mesothelial barrier, is a critical regulator in many cancers. It, however, is still unknown whether miR-23a regulates pancreatic cell proliferation and apoptosis or not. We sought to investigate the role of miR-23a in regulation of pancreatic cell proliferation and apoptosis. miRNA, mRNA, and protein expressions were determined by qRT-PCR and Western blot, respectively. Dual-luciferase reporter assay was used in detection for binding ability of miR-23a to APAF1. Ectopic miR-23a and APAF 1 were introduced to pancreatic cells, and their roles in proliferation and apoptosis were detected by MTT, colony formation, and apoptosis assays, respectively. Up-regulation of miR-23a and down-regulation of APAF 1 were found in pancreatic ductal cancer, respectively. miR-23a significantly inhibited the luciferase activity by targeting APAF 1 3'UTR. Ectopic miR-23a significantly suppressed the APAF 1 gene expression in pancreatic cancer cells. Similar to siAPAF1, miR-23a significantly promoted pancreatic cancer cell proliferation and repressed apoptosis. Furthermore, miR-23a inhibitor and exogenous APAF 1 could recover the effects. It is suggested that miR-23a, acting as an oncogenic regulator by directly targeting APAF 1 in pancreatic cancer, is a useful potential biomarker in diagnosis and treatment of pancreatic cancer.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)