Merck
  • Boronic prodrug of 4-hydroxytamoxifen is more efficacious than tamoxifen with enhanced bioavailability independent of CYP2D6 status.
Boronic prodrug of 4-hydroxytamoxifen is more efficacious than tamoxifen with enhanced bioavailability independent of CYP2D6 status.
BMC cancer (2015-09-12)
Qiu Zhong, Changde Zhang, Qiang Zhang, Lucio Miele, Shilong Zheng, Guangdi Wang
ABSTRACT
Poor initial response to tamoxifen due to CYP2D6 polymorphism and adverse side effects are two clinical challenges in tamoxifen therapy. We report the development and preclinical testing of a boronic prodrug to orally deliver 4-OHT at therapeutically effective concentrations but at a fraction of the standard tamoxifen dose. A mouse xenograft tumor model was used to investigate the efficacy of ZB497 in comparison with tamoxifen. Pharmacokinetic studies were conducted to evaluate the metabolism and bioavailability of the drug in mice. Drug and metabolites distribution in xenograft tumor tissues was determined by high performance liquid chromatography-tandem mass spectrometry. The boronic prodrug, ZB497, can not only be efficiently converted to 4-OHT in mice, but also afforded over 30 fold higher plasma concentrations of 4-OHT than in mice given either the same dose of 4-OHT or tamoxifen. Further, ZB497 was more effective than tamoxifen at lowered dosage in inhibiting the growth of xenograft tumors in mice. Consistent with these observations, ZB497 treated mice accumulated over 6 times higher total drug concentrations than tamoxifen treated mice. Our study demonstrates that ZB497 effectively delivers a markedly increased plasma concentration of 4-OHT in mice. The boronic prodrug was shown to have far superior bioavailability of 4-OHT compared to tamoxifen or 4-OHT administration as measured by the area under the plasma concentration time curve (AUC), plasma peak concentrations, and drug accumulation in tumor tissues. Further, ZB497 proves to be a more efficacious hormone therapy than tamoxifen administered at a reduced dose in mice.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC