Merck
  • Strong positive cooperativity in binding to the A3T3 repeat by Hoechst 33258 derivatives attaching the quinoline units at the end of a branched linker.
Strong positive cooperativity in binding to the A3T3 repeat by Hoechst 33258 derivatives attaching the quinoline units at the end of a branched linker.
Bioorganic & medicinal chemistry (2015-07-15)
Hironori Koda, John Alan Brazier, Ippei Onishi, Shigeki Sasaki
ABSTRACT
Hoechst 33258 derivatives with additional interacting moieties attached at the ends of branched linkers were synthesized, and their DNA binding properties were investigated with regard to the A3T3 repeat by measuring fluorescence spectra. The binding property of the ligand was investigated by fluorescence titration, and the titration data were analyzed using the McGhee-von Hippel method. Ligand 6Q with the quinolin-6-yloxyacetyl group and Ligand IQ with isoquinolin-6-yloxyacetyl group at the ends of the branched linkers exhibit highly positive cooperativity for the DNA having 5 A3T3 sites with 3 base-insertions between them with sequence selectivity. The strategy developed in this study may be generally applicable for designing ligands for repetitive DNA sequences.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Molecular Biology, sterile filtered