Merck
  • Cryptotanshinone Reverses Cisplatin Resistance of Human Lung Carcinoma A549 Cells through Down-Regulating Nrf2 Pathway.
Cryptotanshinone Reverses Cisplatin Resistance of Human Lung Carcinoma A549 Cells through Down-Regulating Nrf2 Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-09-12)
Chen Xia, Xupeng Bai, Xiangyu Hou, Xiaoli Gou, Yongtao Wang, Hang Zeng, Min Huang, Jing Jin
ABSTRACT
To explore whether Nrf2 was associated with drug-resistance in cisplatin resistant A549 (A549/DDP) cells, and if cryptotanshinone (CTS), one of the bioactive compounds isolated from the roots of Salvia miltiorrhiza Bunge (Danshen), could enhance the sensitivity in A549/DDP cells towards cisplatin. A549 and A549/DDP cells were subjected to various treatments, and then Sulforhodamine B (SRB) assay, flow cytometry analysis and western immunoblotting analysis were applied to determine IC50, apoptotic status and expressions of Nrf2 and its downstream genes. The endogenous expression levels of Nrf2 as well as its target genes including GCLC, GCLM, HO-1, NQO1 and MRP1 were much higher in A549/DDP cells than those of A549 cells and the susceptibility of A549/DDP cells to cisplatin was partially restored by silencing Nrf2. The combination of CTS and cisplatin led to cell death and apoptosis through sensitizing A549/DDP cells towards cisplatin compared with cisplatin mono-treatment, however, this reversal role could be abolished by Nrf2 knockdown. Specifically, CTS obviously diminished Nrf2 expression, thus contributing to the decrease of Nrf2-target genes expression levels. Meanwhile, we also discovered that CTS triggered several other signals involving in chemoresistance such as MAPKs, Akt and STAT3 pathway. Our data indicated CTS may be developed as a potential sensitizer cooperating with anticancer drugs to combat chemoresistant carcinoma through the inhibition of the Nrf2 pathway.
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis