- Multi-Biomarkers for Early Detection of Type 2 Diabetes, Including 10- and 12-(Z,E)-Hydroxyoctadecadienoic Acids, Insulin, Leptin, and Adiponectin.
Multi-Biomarkers for Early Detection of Type 2 Diabetes, Including 10- and 12-(Z,E)-Hydroxyoctadecadienoic Acids, Insulin, Leptin, and Adiponectin.
We have previously found that fasting plasma levels of totally assessed 10- and 12-(Z,E)-hydroxyoctadecadienoic acid (HODE) correlated well with levels of glycated hemoglobin (HbA1c) and glucose during oral glucose tolerance tests (OGTT); these levels were determined via liquid chromatography-mass spectrometry after reduction and saponification. However, 10- and 12-(Z,E)-HODE alone cannot perfectly detect early impaired glucose tolerance (IGT) and/or insulin resistance, which ultimately lead to diabetes. In this study, we randomly recruited healthy volunteers (n = 57) who had no known history of any diseases, and who were evaluated using the OGTT, the HODE biomarkers, and several additional proposed biomarkers, including retinol binding protein 4 (RBP4), adiponectin, leptin, insulin, glycoalbumin, and high sensitivity-C-reactive protein. The OGTT revealed that our volunteers included normal individuals (n = 44; Group N), "high-normal" individuals (fasting plasma glucose 100-109 mg/dL) with IGT (n = 11; Group HN+IGT), and diabetic individuals (n = 2; Group D). We then used these groups to evaluate the potential biomarkers for the early detection of type 2 diabetes. Plasma levels of RBP4 and glycoalbumin were higher in Group HN+IGT, compared to those in Group N, and fasting levels of 10- and 12-(Z,E)-HODE/linoleic acids were significantly correlated with levels of RBP4 (p = 0.003, r = 0.380) and glycoalbumin (p = 0.006, r = 0.316). Furthermore, we developed a stepwise multiple linear regression models to predict the individuals' insulin resistance index (the Matsuda Index 3). Fasting plasma levels of 10- and 12-(Z,E)-HODE/linoleic acids, glucose, insulin, and leptin/adiponectin were selected as the explanatory variables for the models. The risks of type 2 diabetes, early IGT, and insulin resistance were perfectly predicted by comparing fasting glucose levels to the estimated Matsuda Index 3 (fasting levels of 10- and 12-(Z,E)-HODE/linoleic acids, insulin, and leptin/adiponectin).