Merck
  • Stereoselective Determination of Tebuconazole in Water and Zebrafish by Supercritical Fluid Chromatography Tandem Mass Spectrometry.
Stereoselective Determination of Tebuconazole in Water and Zebrafish by Supercritical Fluid Chromatography Tandem Mass Spectrometry.
Journal of agricultural and food chemistry (2015-07-01)
Na Liu, Fengshou Dong, Jun Xu, Xingang Liu, Zenglong Chen, Yan Tao, Xinglu Pan, XiXi Chen, Yongquan Zheng
ABSTRACT
A simple and sensitive method for the enantioselective determination of tebuconazole enantiomers in water and zebrafish has been established using supercritical fluid chromatography (SFC)-MS/MS. The effects of the chiral stationary phases, mobile phase, auto back pressure regulator (ABPR) pressure, column temperature, flow rate of the mobile phase, and compensation pump solvent were evaluated. Finally, the optimal SFC-MS/MS working conditions were determined to include a CO2/MeOH mobile phase (87:13, v/v), 2.0 mL/min flow rate, 2200 psi ABPR, and 30 °C column temperature using a Chiralpak IA-3 chiral column under electrospray ionization positive mode. The modified QuEChERS method was applied to water and zebrafish samples. The mean recoveries for the tebuconazole enantiomers were 79.8-108.4% with RSDs ≤ 7.0% in both matrices. The LOQs ranged from 0.24 to 1.20 μg/kg. The developed analytical method was further validated by application to the analysis of authentic samples.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles