Merck
  • Direct conversion of human fibroblasts into functional osteoblasts by defined factors.
Direct conversion of human fibroblasts into functional osteoblasts by defined factors.
Proceedings of the National Academy of Sciences of the United States of America (2015-04-29)
Kenta Yamamoto, Tsunao Kishida, Yoshiki Sato, Keisuke Nishioka, Akika Ejima, Hiroyoshi Fujiwara, Toshikazu Kubo, Toshiro Yamamoto, Narisato Kanamura, Osam Mazda
ABSTRACT
Osteoblasts produce calcified bone matrix and contribute to bone formation and remodeling. In this study, we established a procedure to directly convert human fibroblasts into osteoblasts by transducing some defined factors and culturing in osteogenic medium. Osteoblast-specific transcription factors, Runt-related transcription factor 2 (Runx2), and Osterix, in combination with Octamer-binding transcription factor 3/4 (Oct4) and L-Myc (RXOL) transduction, converted ∼ 80% of the fibroblasts into osteocalcin-producing cells. The directly converted osteoblasts (dOBs) induced by RXOL displayed a similar gene expression profile as normal human osteoblasts and contributed to bone repair after transplantation into immunodeficient mice at artificial bone defect lesions. The dOBs expressed endogenous Runx2 and Osterix, and did not require continuous expression of the exogenous genes to maintain their phenotype. Another combination, Oct4 plus L-Myc (OL), also induced fibroblasts to produce bone matrix, but the OL-transduced cells did not express Osterix and exhibited a more distant gene expression profile to osteoblasts compared with RXOL-transduced cells. These findings strongly suggest successful direct reprogramming of fibroblasts into functional osteoblasts by RXOL, a technology that may provide bone regeneration therapy against bone disorders.
Sigma-Aldrich
Allene, ≥95%