Merck
  • HIF-2 Inhibition Supresses Inflammatory Responses and Osteoclastic Differentiation in Human Periodontal Ligament Cells.
HIF-2 Inhibition Supresses Inflammatory Responses and Osteoclastic Differentiation in Human Periodontal Ligament Cells.
Journal of cellular biochemistry (2015-01-08)
Won-Jung Bae, Mee-Ran Shin, Soo-Kyung Kang, Zhang-Jun, Jun-Yeol Kim, Sang-Cheon Lee, Eun-Cheol Kim
ABSTRACT
Recent reports suggest that hypoxia inducible factor-2α (HIF-2α) is a key regulator of osteoarthritis cartilage destruction. However, the precise role of HIF-2α in the inflammatory response and osteoclast differentiation remains unclear. The purpose of this study was to investigate the effect of HIF-2α on inflammatory cytokines, extracellular matrix (ECM) destruction enzymes, and osteoclastic differentiation in nicotine and lipopolysaccharide (LPS)-stimulated human periodontal ligament cells (PDLCs). HIF-2α was upregulated in chronically inflamed PDLCs of periodontitis patients, and in nicotine- and LPS-exposed PDLC in dose- and time-dependent manners. HIF-2α inhibitor and HIF-2α siRNA attenuated the nicotine- and LPS- induced production of NO and PGE2 , upregulation of iNOS, COX-2, pro-inflammatory cytokines (IL-1β, TNF-α, IL-1β, IL-6, IL-8, IL-10, IL-11, and IL-17), and matrix metalloproteinases (MMPs; MMP-1, -8, -13, -2 and -9), and reversed the effect on TIMPs (TIMP-1 and -2) in PDLCs. The conditioned medium produced by nicotine and LPS-treated PDLCs increased the number of TRAP-stained osteoclasts, TRAP activity and osteoclast-specific genes, which has been blocked by HIF-2α inhibition and silencing. HIF-2α inhibitor and HIF-2α siRNA inhibited the effects of nicotine and LPS on the activation of Akt, JAK2 and STAT3, ERK and JNK MAPK, nuclear factor-κB, c-Jun, and c-Fos. Taken together, this study is the first to demonstrate that HIF-2α inhibition exhibits anti-inflammatory activity through the inhibition of inflammatory cytokines and impairment of ECM destruction, as well as blocking of osteoclastic differentiation in a nicotine- and periodontopathogen-stimulated PDLCs model. Thus, HIF-2α inhibition may be a novel molecular target for therapeutic approaches in periodontitis.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG