Merck
  • Synthesis and evaluation of 1H-pyrrolo[2,3-b]pyridine derivatives as novel immunomodulators targeting Janus kinase 3.
Synthesis and evaluation of 1H-pyrrolo[2,3-b]pyridine derivatives as novel immunomodulators targeting Janus kinase 3.
Chemical & pharmaceutical bulletin (2015-03-20)
Yutaka Nakajima, Takashi Tojo, Masataka Morita, Keiko Hatanaka, Shohei Shirakami, Akira Tanaka, Hiroshi Sasaki, Kazuo Nakai, Koichiro Mukoyoshi, Hisao Hamaguchi, Fumie Takahashi, Ayako Moritomo, Yasuyuki Higashi, Takayuki Inoue
ABSTRACT
Janus kinases (JAKs) have been known to play crucial roles in modulating a number of inflammatory and immune mediators. Here, we describe a series of 1H-pyrrolo[2,3-b]pyridine derivatives as novel immunomodulators targeting JAK3 for use in treating immune diseases such as organ transplantation. In the chemical modification of compound 6, the introduction of a carbamoyl group to the C5-position and substitution of a cyclohexylamino group at the C4-position of the 1H-pyrrolo[2,3-b]pyridine ring led to a large increase in JAK3 inhibitory activity. Compound 14c was identified as a potent, moderately selective JAK3 inhibitor, and the immunomodulating effect of 14c on interleukin-2-stimulated T cell proliferation was shown. Docking calculations and WaterMap analysis of the 1H-pyrrolo[2,3-b]pyridine-5-carboxamide derivatives were conducted to confirm the substituent effects on JAK3 inhibitory activity.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis