Merck
  • An 8-year longitudinal study of overreaching in 114 elite female Chinese wrestlers.
An 8-year longitudinal study of overreaching in 114 elite female Chinese wrestlers.
Journal of athletic training (2014-10-21)
Ye Tian, Zihong He, Jiexiu Zhao, Dalang Tao, Kuiyuan Xu, Adrian Midgley, Lars McNaughton
ABSTRACT
Successful training involves structured overload but must avoid the combination of excessive overload and inadequate recovery. The aim of this study was to determine the incidence of functional overreaching (FOR), nonfunctional overreaching (NFOR), and overtraining syndrome in elite female wrestlers during their normal training and competition schedules and to explore the utility of blood markers for the early detection of overreaching. Classification of FOR, NFOR, and overtraining syndrome was based on the European Congress of Sports Medicine position statement. Case series. China Institute of Sport Science. Over an 8-year period, 114 wrestlers from the women's Asian wrestling team were monitored to help identify if and when they experienced FOR, NFOR, or overtraining syndrome. Creatine kinase, hemoglobin, testosterone, and cortisol were measured throughout the period to identify whether wrestlers were outside the reference intervals (constructed from normal recovery data) during periods of overreaching and not overreaching. Among the 114 athletes, there were 13 (3.6%) instances of FOR, 23 (6.4%) instances of NFOR, and 2 (0.6%) instances of overtraining syndrome. The diagnostic sensitivity for FOR was 38%, 15%, 45%, and 18% for creatine kinase, hemoglobin, testosterone, and cortisol, respectively. The diagnostic sensitivity for NFOR was 29%, 33%, 26%, and 35% for creatine kinase, hemoglobin, testosterone, and cortisol, respectively. Specificity was 79%, 88%, 90%, and 82% for creatine kinase, hemoglobin, testosterone, and cortisol, respectively. Post hoc analysis showed no mean differences in creatine kinase (F = 0.5, P = .47), hemoglobin (F = 3.8, P = .052), testosterone (F = 0.2, P = .62), or cortisol (F = 0.04, P = .85) between monitoring periods when wrestlers were and were not diagnosed with FOR and NFOR. Coaches and sports scientists should not use single blood variables as markers of overreaching in elite female wrestlers.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Testosterone, ≥98%
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®