Merck
  • The effects of atorvastatin on the prevention of osteoporosis and dyslipidemia in the high-fat-fed ovariectomized rats.
The effects of atorvastatin on the prevention of osteoporosis and dyslipidemia in the high-fat-fed ovariectomized rats.
Calcified tissue international (2015-03-31)
Sien Lin, Jianping Huang, Ziwei Fu, Yanlong Liang, Haiyou Wu, Liangliang Xu, Yuxin Sun, Wayne Y W Lee, Tie Wu, Ling Qin, Liao Cui, Gang Li
ABSTRACT
Previous studies reported that statins showed positive effects on bone in both human and animal models. This study aimed to investigate the effects of atorvastatin on the prevention of osteoporosis and dyslipidemia in ovariectomized rats fed with high-fat emulsion. The 3-month-old female rats were subjected to either sham operations (n = 8) or ovariectomized operations (OVX, n = 24). The OVX rats were orally administered deionized water (n = 8) or standardized high-fat emulsion without (n = 8) or with atorvastatin (n = 8). All rats were injected twice with calcein before sacrificed for the purpose of double in vivo labeling. After 12 weeks, all rats were sacrificed under anesthesia. Biochemistry, histomorphometry, mechanical test, micro-computed tomography analysis, mechanical test, histology, and component analysis were performed. We found that high-fat emulsion significantly decreased body weight, bone formation, collagen content of bone, and bone biomechanics, while increased blood, liver, and bone marrow lipids. Atorvastatin treatment prevented dyslipidemia, reversed hepatic steatosis, optimized composition of bone, and improved bone mechanical properties. The current study provided further evidence that atorvastatin might be useful for the treatment of osteoporotic patients with dyslipidemia.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications