Merck
  • Double Sonogashira reactions on dihalogenated aminopyridines for the assembly of an array of 7-azaindoles bearing triazole and quinoxaline substituents at C-5: Inhibitory bioactivity against Giardia duodenalis trophozoites.
Double Sonogashira reactions on dihalogenated aminopyridines for the assembly of an array of 7-azaindoles bearing triazole and quinoxaline substituents at C-5: Inhibitory bioactivity against Giardia duodenalis trophozoites.
Bioorganic & medicinal chemistry (2015-06-06)
Tlabo C Leboho, Somnath Giri, Inessa Popova, Ian Cock, Joseph P Michael, Charles B de Koning
ABSTRACT
The synthesis of 2,3,5-trisubstituted 7-azaindoles as well as 2,5-disubstituted 7-azaindoles from 3,5-dihalogenated 2-aminopyridines is outlined. Using a double Sonogashira coupling reaction on 2-amino-3,5-diiodopyridine followed by the Cacchi reaction the synthesis of 2,3,5-trisubstituted 7-azaindoles was accomplished. In addition, using two sequential Sonogashira coupling reactions on 2-amino-5-bromo-3-iodopyridine and a potassium t-butoxide mediated ring closure reaction resulted in the assembly of 2,5-disubstituted 7-azaindoles. The 5-alkynyl substituent of the azaindole was easily converted into both quinoxaline and triazole substituents, the latter utilizing an alkyne-azide cycloaddition reaction. Some of these azaindole derivatives showed very promising biological activity against the gastrointestinal protozoal parasite Giardia duodenalis.
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate