Merck
  • A simple model for pollen-parent fecundity distributions in bee-pollinated forage legume polycrosses.
A simple model for pollen-parent fecundity distributions in bee-pollinated forage legume polycrosses.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik (2015-06-25)
Heathcliffe Riday, Mark A Smith, Michael D Peel
ABSTRACT
A simple Weibull distribution based empirical model that predicts pollen-parent fecundity distributions based on polycross size alone has been developed in outbred forage legume species for incorporation into quantitative genetic theory. Random mating or panmixis is a fundamental assumption in quantitative genetic theory. Random mating is sometimes thought to occur in actual fact, although a large body of empirical work shows that this is often not the case in nature. Models have been developed to explain many non-random mating phenomena. This paper measured pollen-parent fecundity distributions among outbred perennial forage legume species [autotetraploid alfalfa (Medicago sativa L.), autohexaploid kura clover (Trifolium ambiguum M. Bieb.), and diploid red clover (Trifolium pratense L.)] in ten polycrosses ranging in size (N) from 9 to 94 pollinated with bee pollinators [Bumble Bees (Bombus impatiens Cr.) and leafcutter bees (Megachile rotundata F.)]. A Weibull distribution best fit the observed pollen-parent fecundity distributions. After standardizing data among the 10 polycrosses, a single Weibull distribution-based model was obtained with an R (2) of 0.978. The model is able to predict pollen-parent fecundity distributions based on polycross size alone. The model predicts that the effective polycross size will be approximately 9 % smaller than under random mating (i.e., N e/N ~ 0.91). The model is simple and can easily be incorporated into other models or simulations requiring a pollen-parent fecundity distribution. Further work is needed to determine how widely applicable the model is.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology