Merck
  • Choline concentrations are lower in postnatal plasma of preterm infants than in cord plasma.
Choline concentrations are lower in postnatal plasma of preterm infants than in cord plasma.
European journal of nutrition (2014-08-26)
Wolfgang Bernhard, Marco Raith, Rebecca Kunze, Vera Koch, Martin Heni, Christoph Maas, Harald Abele, Christian F Poets, Axel R Franz
ABSTRACT
Choline is essential to human development, particularly of the brain in the form of phosphatidylcholine, sphingomyelin and acetylcholine, for bile and lipoprotein formation, and as a methyl group donator. Choline is actively transported into the fetus, and maternal supply correlates with cognitive outcome. Interruption of placental supply may therefore impair choline homeostasis in preterm infants. Determination of postnatal plasma concentrations of choline and its derivatives betaine and dimethylglycine (DMG) in preterm infants compared to cord and maternal blood matched for postmenstrual age (PMA). We collected plasma of very low-birth-weight infants undergoing neonatal intensive care (n = 162), cord plasma of term and preterm infants (n = 176, 24-42-week PMA), serum of parturients (n = 36), and plasma of healthy premenopausal women (n = 40). Target metabolites were analyzed with tandem mass spectrometry and reported as median (25th/75th percentiles). Cord plasma choline concentration was 41.4 (31.8-51.2) µmol/L and inversely correlated with PMA. In term but not in preterm infants, cord plasma choline was lower in girls than in boys. Prenatal glucocorticoid treatment did not affect choline levels in cord plasma, whereas betaine was decreased and DMG increased. In parturients and non-pregnant women, choline concentrations were 14.1 (10.3-16.9) and 8.8 (5.7-11.2) µmol/L, respectively, whereas betaine was lowest in parturients. After delivery, preterm infant plasma choline decreased to 20.8 (16.0-27.6) µmol/L within 48 h. Betaine and DMG correlated with plasma choline in all groups. In preterm infants, plasma choline decreases to 50 % of cord plasma concentrations, reflecting choline undernourishment and postnatal metabolic adaptation, and potentially contributing to impaired outcome.
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
N,N-Dimethylglycine, ≥99%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer