Merck
  • Identification of Echinacoside Metabolites Produced by Human Intestinal Bacteria Using Ultraperformance Liquid Chromatography-Quadrupole Time-of-Flight Mass Spectrometry.
Identification of Echinacoside Metabolites Produced by Human Intestinal Bacteria Using Ultraperformance Liquid Chromatography-Quadrupole Time-of-Flight Mass Spectrometry.
Journal of agricultural and food chemistry (2015-07-18)
Yang Li, Guisheng Zhou, Shihua Xing, Pengfei Tu, Xiaobo Li
ABSTRACT
Echinacoside (ECH) is one of the representative phenylethanoid glycosides. It is widely present in plants and exhibits various bioactivities. However, the extremely low oral bioavailability of ECH in rats implies that ECH may go through multiple hydrolysis steps in the gastrointestinal tract prior to its absorption into the blood. Therefore, the gastrointestinal metabolites of ECH are more likely to be the bioactive components. This study established an approach combining ultraperformance liquid chromatography-quadrupole time-of-flight mass spectrometry (UPLC-Q-TOF-MS) with MS(E) technology and MetaboLynx software for rapid analysis of the ECH metabolic profile produced by human intestinal bacteria. As a result, 13 ECH metabolites and 5 possible metabolic pathways (including deglycosylation, dehydroxylation, reduction, hydroxylation, and acetylation) were identified. Furthermore, hydroxytyrosol (HT) and 3-hydroxyphenylpropionic acid (3-HPP) were found to be the two bioactive metabolites of ECH produced by human intestinal bacteria.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Hydrocinnamic acid, 99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC