Merck
  • Characterization of a novel thermostable patatin-like protein from a Guaymas basin metagenomic library.
Characterization of a novel thermostable patatin-like protein from a Guaymas basin metagenomic library.
Extremophiles : life under extreme conditions (2015-05-29)
Ling Fu, Ying He, Fangdi Xu, Qun Ma, Fengping Wang, Jun Xu
ABSTRACT
Deep-sea hydrothermal vents are a natural habitat for thermophiles, in which contain plenty of enzymes that can function at high temperatures. In this work, we constructed a fosmid library in Escherichia coli using metagenomic DNA isolated from a chimney sample collected in the hydrothermal vents in Guaymas Basin. The library was screened for lipolytic activity and positive clones were subjected to subcloning. A novel patatin-like protein (PLP) that exhibited less than 45 % identity in amino acid sequence to known enzymes was obtained. Common features of the patatin-like proteins, such as four conserved blocks, were detected. Interestingly, there was an Ala at site 42 in PLP instead of the first Gly-residue in the consensus sequence Gly-X-Ser-X-Gly found in other PLP homologs. The active sites of PLP were Ser44 and Asp160. Spectrophotometric assays with different p-nitrophenyl esters demonstrated a preference for p-nitrophenyl butyrate (C4) and p-nitrophenyl decanoate (C10). Moreover, PLP demonstrated optimal activity at 70 °C and at pH 9.0 (Tris-HCl). The activation energy from the linear Arrhenius plot was found to be 38.3 ± 0.9 kJ/mol. The K m and V max of PLP for C4 were 304 ± 38 μM and 14 ± 0.38 μmol min(-1) mg(-1), respectively. Gene-mining of the metagenome dataset that was generated by pyrosequencing the same chimney sample resulted in identification of 20 PLP homolog gene fragments, which could represent promising examples of this category of thermostable proteins.
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder