Merck
  • Silencing expression of the clusterin/apolipoprotein j gene in human cancer cells using small interfering RNA induces spontaneous apoptosis, reduced growth ability, and cell sensitization to genotoxic and oxidative stress.
Silencing expression of the clusterin/apolipoprotein j gene in human cancer cells using small interfering RNA induces spontaneous apoptosis, reduced growth ability, and cell sensitization to genotoxic and oxidative stress.
Cancer research (2004-03-05)
Ioannis P Trougakos, Alan So, Burkhard Jansen, Martin E Gleave, Efstathios S Gonos
ABSTRACT
Clusterin/Apolipoprotein J (CLU) is a heterodimeric ubiquitously expressed secreted glycoprotein that is implicated in several physiological processes and is differentially expressed in many severe physiological disturbances, including tumor formation and in vivo cancer progression. Despite extensive efforts, clarification of CLU's biological role has been exceptionally difficult and its precise function remains elusive. Short RNA duplexes, referred to as small interfering RNAs (siRNAs), provide a new approach for the elucidation of gene function in human cells. Here, we describe siRNA-mediated CLU gene silencing in osteosarcoma and prostate human cancer cells and illustrate that CLU mRNA is amenable to siRNA-mediated degradation. Our data demonstrate that CLU knockdown in human cancer cells induces significant reduction of cellular growth and higher rates of spontaneous endogenous apoptosis. Moreover, CLU knockdown cancer cells were significantly sensitized to both genotoxic and oxidative stress induced by chemotherapeutic drugs and H(2)O(2), respectively. These effects were more pronounced in cell lines that express high endogenous steady-state levels of the CLU protein and occur through hyperactivation of the cellular apoptotic machinery. Overall, our results reveal that, in the distinct cellular contexts of the osteosarcoma and prostate cancer cells assayed, CLU is a central molecule in cell homeostasis that exerts a cytoprotective function. The described CLU-specific siRNA oligonucleotides that can potently silence CLU gene expression may thus prove valuable agents during antitumor therapy or at other pathological conditions where CLU has been implicated.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)