Merck
  • Vitamin B1 Deficiency Does not Affect the Liver Concentrations of the Other Seven Kinds of B-Group Vitamins in Rats.
Vitamin B1 Deficiency Does not Affect the Liver Concentrations of the Other Seven Kinds of B-Group Vitamins in Rats.
Nutrition and metabolic insights (2013-08-13)
Katsumi Shibata, Atsushi Shimizu, Tsutomu Fukuwatari
ABSTRACT
We aimed to determine the effects of vitamin B1 deficiency on vitamin contents of urine, liver, and blood. In the current study, rats were divided into 3 groups (n = 5, each group): the first was freely fed a complete diet (ad lib-fed control group); the second freely fed a vitamin B1-free diet (vitamin B1 deficient group); and the third pair-fed a complete diet with the same amounts of the vitamin B1 deficient group (pair-fed control group). The experimental period was for 15 days. The blood concentrations of vitamin B2, PLP, vitamin B12, folic acid, and biotin were lower in the pair-fed control than in the ad lib-fed control and those of nicotinamide and pantothenic acid were the same. We conclude that Vitamin B1 deficiency did not affect concentrations of the other B-group vitamins.
Sigma-Aldrich
4-Pyridoxic acid, ≥98%