Merck
  • In vivo effects of Faizol Ubat Batuk, a herbal product on aminopyrine metabolism in rat hepatocytes.
In vivo effects of Faizol Ubat Batuk, a herbal product on aminopyrine metabolism in rat hepatocytes.
The Libyan journal of medicine (2011-10-07)
Yousef A Taher, Abas Hj Hussin
ABSTRACT
Traditional medicines, in particular herbal products, have been used abundantly over the years in curing several diseases. Pharmacological interactions of herbal products with modern drugs, however, remain to some extent unknown. Herein, we examined whether co-administration of Faizol Ubat Batuk (FUB), a mixture of aqueous extract of different plants, modifies the metabolism of aminopyrine, a conventional analgesic drug, in rat liver. We used rat hepatocytes outfitted by collagenase perfusion technique. Determination of aminopyrine n-demethylase activity was performed using the Nash colorimetric method, by measuring the amount of formaldehyde produced. Compared to control treatment, FUB significantly increased the hepatic metabolism of aminopyrine in healthy adult male rats. In contrast, the hepatic metabolism of aminopyrine in adult female rats was decreased. Besides, a biphasic effect in n-demethylase activity was observed in young male rats treated with FUB. In a subsequent experiment, FUB did not change the metabolism of aminopyrine in streptozotocin (STZ)-diabetic adult male rats. In conclusion, administration of FUB could affect phase I aminopyrine metabolism in rat heptocytes. In addition, the effects of FUB on hepatic n-demethylase activity were gender and disease dependent.
Sigma-Aldrich
4-Dimethylaminoantipyrine, reactive nitrogen species scavenger