Merck
  • A continuous fluorescent assay for the determination of plasma and tissue angiotensin I-converting enzyme activity.
A continuous fluorescent assay for the determination of plasma and tissue angiotensin I-converting enzyme activity.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas (2005-06-04)
M F Alves, M C Araujo, M A Juliano, E M Oliveira, J E Krieger, D E Casarini, L Juliano, A K Carmona
ABSTRACT
A continuous assay using internally quenched fluorescent peptides with the general sequence Abz-peptidyl-(Dnp)P-OH (Abz = ortho-aminobenzoic acid; Dnp = 2,4-dinitrophenyl) was optimized for the measurement of angiotensin I-converting enzyme (ACE) in human plasma and rat tissues. Abz-FRK(Dnp)P-OH, which was cleaved at the Arg-Lys bond by ACE, was used for the enzyme evaluation in human plasma. Enzymatic activity was monitored by continuous recording of the fluorescence (lambda ex = 320 nm and lambda em = 420 nm) at 37 degrees C, in 0.1 M Tris-HCl buffer, pH 7.0, with 50 mM NaCl and 10 microM ZnCl2. The assays can be performed directly in the cuvette of the fluorimeter and the hydrolysis followed for 5 to 10 min. ACE measurements in the plasma of 80 healthy patients with Hip-His-Leu and with Abz-FRK(Dnp)P-OH correlated closely (r = 0.90, P < 0.001). The specificity of the assay was demonstrated by the complete inhibition of hydrolysis by 0.5 microM lisinopril or captopril. Abz-FRK(Dnp)P-OH cleavage by ACE was monitored in rat lung, kidney, heart, and liver homogenates in the presence of a cocktail of inhibitors containing trans-epoxy-succinyl-L-leucylamido-(4-guanido)-butene, pepstatin, phenyl-methylsulfonyl fluoride, N-tosyl-L-phenylalanyl-chloromethyl ketone, and N-tosyl-lysyl-chloromethyl ketone to prevent undesirable hydrolysis. ACE activity in lung, heart and kidney homogenates, but not in liver homogenates, was completely abolished by 0.5 microM lisinopril or captopril. The advantages of the method are the procedural simplicity and the high sensitivity providing a rapid assay for ACE determinations.
Sigma-Aldrich
Anthranilic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Anthranilic acid, reagent grade, ≥98%
Sigma-Aldrich
Abz-FRK(Dnp)P-OH trifluoroacetate salt, ≥95% (HPLC), film