Merck
Total synthesis of (+)-macquarimicin A.
Journal of the American Chemical Society (2003-12-04)
Ryosuke Munakata, Hironori Katakai, Tatsuo Ueki, Jun Kurosaka, Ken-ichi Takao, Kin-ichi Tadano
ABSTRACT
The first total synthesis of (+)-macquarimicin A (1), a novel inhibitor of neutral sphingomyelinase (N-SMase) with antiinflammatory activity, has been accomplished. The present work determined the absolute configuration of (+)-1 and revised the C(2)-C(3) geometry to be Z. The synthesis features a transannular Diels-Alder reaction, which constructed the tetracyclic framework stereoselectively, and a convergent and efficient synthetic pathway, which afforded (+)-macquarimicin A (1) in 27 steps (longest linear sequence) with 9.9% overall yield.
Sigma-Aldrich
(S)-(+)-Epichlorohydrin, 98%
Pricing and availability is not currently available.