Merck
  • Hedgehog associated to microparticles inhibits adipocyte differentiation via a non-canonical pathway.
Hedgehog associated to microparticles inhibits adipocyte differentiation via a non-canonical pathway.
Scientific reports (2016-03-25)
Audrey Fleury, Lucile Hoch, M Carmen Martinez, Hélène Faure, Maurizio Taddei, Elena Petricci, Fabrizio Manetti, Nicolas Girard, André Mann, Caroline Jacques, Jérôme Larghero, Martial Ruat, Ramaroson Andriantsitohaina, Soazig Le Lay
ABSTRACT
Hedgehog (Hh) is a critical regulator of adipogenesis. Extracellular vesicles are natural Hh carriers, as illustrated by activated/apoptotic lymphocytes specifically shedding microparticles (MP) bearing the morphogen (MP(Hh+)). We show that MP(Hh+) inhibit adipocyte differentiation and orientate mesenchymal stem cells towards a pro-osteogenic program. Despite a Smoothened (Smo)-dependency, MP(Hh+) anti-adipogenic effects do not activate a canonical Hh signalling pathway in contrast to those elicited either by the Smo agonist SAG or recombinant Sonic Hedgehog. The Smo agonist GSA-10 recapitulates many of the hallmarks of MP(Hh+) anti-adipogenic effects. The adipogenesis blockade induced by MP(Hh+) and GSA-10 was abolished by the Smo antagonist LDE225. We further elucidate a Smo/Lkb1/Ampk axis as the non-canonical Hh pathway used by MP(Hh+) and GSA-10 to inhibit adipocyte differentiation. Our results highlight for the first time the ability of Hh-enriched MP to signal via a non-canonical pathway opening new perspectives to modulate fat development.
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)