Merck
  • Characterization of the autoantigen La (SS-B) as a dsRNA unwinding enzyme.
Characterization of the autoantigen La (SS-B) as a dsRNA unwinding enzyme.
Nucleic acids research (1997-01-15)
P Hühn, G J Pruijn, W J van Venrooij, M Bachmann
ABSTRACT
During the analysis of the La (SS-B) autoantigen for catalytic activities an ATP-dependent double-stranded RNA unwinding activity was detected. Both native and recombinant La proteins from different species displayed this activity, which could be inhibited by monospecific anti-La antibodies. La protein was able to melt dsRNA substrates with either two 3'-overhangs or a single 3'- and a 5'-overhang. Double-stranded RNAs with two 5'-overhangs were not unwound, indicating that at least one 3'-overhang is required for unwinding. Sequence elements of the La protein that might be involved in dsRNA unwinding, such as an evolutionarily conserved putative ATP-binding motif and an element that is homologous to the double-stranded RNA binding protein kinase PKR, are discussed.
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.