Merck
  • Omega-3 polyunsaturated fatty acids critically regulate behaviour and gut microbiota development in adolescence and adulthood.
Omega-3 polyunsaturated fatty acids critically regulate behaviour and gut microbiota development in adolescence and adulthood.
Brain, behavior, and immunity (2016-07-18)
Ruairi C Robertson, Clara Seira Oriach, Kiera Murphy, Gerard M Moloney, John F Cryan, Timothy G Dinan, R Paul Ross, Catherine Stanton
ABSTRACT
Neurodevelopment is strongly influenced by maternal and early-postnatal diet. Omega-3 polyunsaturated fatty acids (n-3 PUFA) are vital structural and functional components of the developing brain. The gut microbiota is also influenced by n-3 PUFA status, however, little is known about the role of maternal and early-life n-3 PUFA intake on offspring gut microbiota development and subsequent interactions with central nervous system functioning and behavioural outcomes. Pregnant female C57BL/6 mice and their male offspring were fed a control (CON), omega-3 deficient (O3-) or omega-3 supplemented (O3+) diet. Cognitive, depressive and social behaviours were assessed through a battery of behaviour tests in the male offspring at both adolescence (week 4-5) and adulthood (week 11-13). Hypothalamic-pituitary-adrenal axis (HPA) activation was assessed by analysis of stress-induced corticosterone production. Fecal microbiota composition was analysed by 16S sequencing at both adolescent and adulthood. In addition, stimulated spleen cytokine levels were assessed. n-3 PUFA interventions induced subtle changes in offspring early-life and adolescent behaviours, which were further evident in adulthood, such that O3- animals displayed impaired communication, social and depression-related behaviours and O3+ animals displayed enhanced cognition. O3- mice displayed an elevated Firmicutes:Bacteroidetes ratio and blunted systemic LPS responsiveness. Contrastingly, O3+ mice displayed greater fecal Bifidobacterium and Lactobacillus abundance and dampened HPA-axis activity. Neurobehavioural development related to cognitive, anxiety and social behaviours, is highly dependent upon in utero and lifelong n-3 PUFA availability. In addition, neurobehavioural changes induced by altering n-3 PUFA status are closely associated with comprehensive alterations in gut microbiota composition, HPA-axis activity and inflammation.
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture