Merck
  • Degradation of the Mitotic Cyclin Clb3 Is not Required for Mitotic Exit but Is Necessary for G1 Cyclin Control of the Succeeding Cell Cycle.
Degradation of the Mitotic Cyclin Clb3 Is not Required for Mitotic Exit but Is Necessary for G1 Cyclin Control of the Succeeding Cell Cycle.
Genetics (2016-10-30)
Kresti Pecani, Frederick R Cross
ABSTRACT
B-type cyclins promote mitotic entry and inhibit mitotic exit. In Saccharomyces cerevisiae, four B-type cyclins, Clb1-4, carry out essential mitotic roles, with substantial but incomplete overlap of function among them. Previous work in many organisms has indicated that B-type cyclin-dependent inhibition of mitotic exit imposes a requirement for mitotic destruction of B-type cyclins. For instance, precise genomic removal of the Clb2 destruction box (D box) prevents mitotic proteolysis of Clb2, and blocks mitotic exit. Here, we show that, despite significant functional overlap between Clb2 and Clb3, D-box-dependent Clb3 proteolysis is completely dispensable for mitotic exit. Removal of the Clb3 D box results in abundant Clb3 protein and associated kinase throughout the cell cycle, but mitotic exit occurs with close to normal timing. Clb3 degradation is required for pre-Start G1 control in the succeeding cell cycle. Deleting the CLB3 D box essentially eliminates all time delay before cell cycle Start following division, even in very small newborn cells. CLB3∆db cells show no cell cycle arrest response to mating pheromone, and CLB3∆db completely bypasses the requirement for CLN G1 cyclins, even in the absence of the early expressed B-type cyclins CLB5,6 Thus, regulated mitotic proteolysis of Clb3 is specifically required to make passage of Start in the succeeding cell cycle "memoryless"-dependent on conditions within that cycle, and independent of events such as B-type cyclin accumulation that occurred in the preceding cycle.
Sigma-Aldrich
Rabbit IgG−Agarose, saline suspension
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution