Merck
  • Salt-inducible kinase induces cytoplasmic histone deacetylase 4 to promote vascular calcification.
Salt-inducible kinase induces cytoplasmic histone deacetylase 4 to promote vascular calcification.
EMBO reports (2017-06-08)
Alon Abend, Omer Shkedi, Michal Fertouk, Lilac H Caspi, Izhak Kehat
ABSTRACT
A pathologic osteochondrogenic differentiation of vascular smooth muscle cells (VSMCs) promotes arterial calcifications, a process associated with significant morbidity and mortality. The molecular pathways promoting this pathology are not completely understood. We studied VSMCs, mouse aortic rings, and human aortic valves and showed here that histone deacetylase 4 (HDAC4) is upregulated early in the calcification process. Gain- and loss-of-function assays demonstrate that HDAC4 is a positive regulator driving this pathology. HDAC4 can shuttle between the nucleus and cytoplasm, but in VSMCs, the cytoplasmic rather than the nuclear activity of HDAC4 promotes calcification, and a nuclear-localized mutant of HDAC4 fails to promote calcification. The cytoplasmic location and function of HDAC4 is controlled by the activity of salt-inducible kinase (SIK). Pharmacologic inhibition of SIK sends HDAC4 to the nucleus and inhibits the calcification process in VSMCs, aortic rings, and
Sigma-Aldrich
o-Cresolphthalein, indicator grade
Sigma-Aldrich
KN-93, ≥98% (HPLC)