Merck
  • Pretreatment of Eucalyptus in biphasic system for furfural production and accelerated enzymatic hydrolysis.
Pretreatment of Eucalyptus in biphasic system for furfural production and accelerated enzymatic hydrolysis.
Bioresource technology (2017-04-23)
Xiudong Zhang, Yuanyuan Bai, Xuefei Cao, Runcang Sun
ABSTRACT
Herein, an efficient biphasic pretreatment process was developed to improve the production of furfural (FF) and glucose from Eucalyptus. The influence of formic acid and NaCl on FF production from xylose in water and various biphasic systems was investigated. Results showed that the addition of formic acid and NaCl significantly promoted the FF yield, and the biphasic system of MIBK (methyl isobutyl ketone)/water exhibited the best performance for FF production. Then the Eucalyptus was pretreated in the MIBK/water system, and a maximum FF yield of 82.0% was achieved at 180°C for 60min. Surface of the pretreated Eucalyptus became relatively rough and loose, and its crystallinity index increased obviously due to the removal of hemicelluloses and lignin. The pretreated Eucalyptus samples showed much higher enzymatic hydrolysis rates (26.2-70.7%) than the raw Eucalyptus (14.5%).
Sigma-Aldrich
2-Methyltetrahydrofuran, BioRenewable, anhydrous, ≥99%, Inhibitor-free