Merck
  • Effects of Chinese herbal medicine Xiangbin prescription on gastrointestinal motility.
Effects of Chinese herbal medicine Xiangbin prescription on gastrointestinal motility.
World journal of gastroenterology (2017-05-20)
Zhi Jiang, Li-Xing Cao, Bo Liu, Qi-Cheng Chen, Wen-Fan Shang, Lu Zhou, Dan-Yan Li, De-An Guo, Zhi-Qiang Chen
ABSTRACT
To investigate the effects of Xiangbin prescription (XBP), a Chinese herbal concoction, on gastrointestinal motility. Forty healthy volunteers were recruited for this randomized controlled trial of XBP. Antroduodenojejunal manometry was used to monitor gastrointestinal motility in these subjects. After the subjects had fasted for at least 12 h, XBP ( Oral administration of XBP significantly increased the amplitude of duodenal contractions [19.5 (13.0-26.7) XBP can stimulate duodenal and jejunal motility and increase the concentrations of plasma motilin and ghrelin. The clinical applicability of XBP in treating GDIM deserves investigation.