Merck
  • Homoserine Kinase from Escherichia coli K-12: Properties,
    Inhibition by L-Threonine, and Regulation of Biosynthesis
Homoserine Kinase from Escherichia coli K-12: Properties,<BR/> Inhibition by L-Threonine, and Regulation of Biosynthesis
Journal of Bacteriology (1974)
J. THEZE