Merck
  • Induction of autophagy by an oleanolic acid derivative, SZC017, promotes ROS-dependent apoptosis through Akt and JAK2/STAT3 signaling pathway in human lung cancer cells.
Induction of autophagy by an oleanolic acid derivative, SZC017, promotes ROS-dependent apoptosis through Akt and JAK2/STAT3 signaling pathway in human lung cancer cells.
Cell biology international (2017-09-08)
Yanlin Song, Lingqi Kong, Bin Sun, Lei Gao, Peng Chu, Anil Ahsan, Eskandar Qaed, Yuan Lin, Jinyong Peng, Xiaodong Ma, Jianbin Zhang, Shisheng Wang, Zeyao Tang
ABSTRACT
The signal transducers and activators of transcription 3 (STAT3) signaling pathway is a common feature in many solid tumors including non-small cell lung cancer, whereas current therapies usually fail to treat this disease in majority of cases. In the present study, we aimed to investigate the cytotoxic effect and the underlying mechanisms of SZC017, an oleanolic acid derivative, on human lung cancer cells. Cell viability was significantly decreased in SZC017-treated lung cancer cells. Mechanistically, SZC017 reduced A549 cell viability by activating both apoptosis and autophagy pathways. SZC017 was able to inhibit the phosphorylation of Akt, JAK2, and STAT3 in A549 cells, resulting in the inactivation of Akt and JAK2/STAT3 signaling pathways. In addition, SZC017 could induce ROS generation and Ca
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%