Merck
  • Quantum-Dot-Based Lateral Flow Immunoassay for Detection of Neonicotinoid Residues in Tea Leaves.
Quantum-Dot-Based Lateral Flow Immunoassay for Detection of Neonicotinoid Residues in Tea Leaves.
Journal of agricultural and food chemistry (2017-10-28)
Shuangjie Wang, Ying Liu, Shasha Jiao, Ying Zhao, Yirong Guo, Mengcen Wang, Guonian Zhu
ABSTRACT
Neonicotinoid insecticides are commonly used for pest control on tea plantations as a result of their broad-spectrum activity. However, neonicotinoid residues released from tea leaves into tea infusions pose a dietary risk to consumers. Therefore, a rapid, sensitive, and reliable on-site detection method for neonicotinoids is needed. We developed a quantum-dot-based fluorescent lateral flow immunochromatographic strip (LFICS) combined with a broad-specific antibody for detection of typical neonicotinoids (imidacloprid, imidaclothiz, and clothianidin), with sensitivities [50% inhibitory concentration (IC
Product Description
Supelco
Thiamethoxam, PESTANAL®, analytical standard
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard
Supelco
Nitenpyram, PESTANAL®, analytical standard
Supelco
Clothianidin, PESTANAL®, analytical standard
Supelco
Thiacloprid, PESTANAL®, analytical standard
Supelco
Dinotefuran, PESTANAL®, analytical standard