Merck
  • Determination of neutral diols and carboxylic acids formed during glycerol electrooxidation by capillary electrophoresis with dual C
Determination of neutral diols and carboxylic acids formed during glycerol electrooxidation by capillary electrophoresis with dual C
Talanta (2017-11-16)
Wanderson Sirley Reis Teixeira, Mauro Sérgio Ferreira Santos, Jonas Gruber, Ivano Gebhardt Rolf Gutz, Fernando Silva Lopes
ABSTRACT
Methods for determination of glycerol and its electrooxidation products (neutral diols and carboxylates) by capillary electrophoresis (CE) with dual capacitively coupled contactless conductivity detectors (C
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
Tartronic acid, ≥97.0%
Sigma-Aldrich
1,3-Dihydroxyacetone dimer, 97%