Merck
  • Engineering complex biological systems in bacteria through recombinase-assisted genome engineering.
Engineering complex biological systems in bacteria through recombinase-assisted genome engineering.
Nature Protocols (2014)
Santos CN and Yoshikuni Y2