Merck
  • Identification of protein lysine methylation readers with a yeast three-hybrid approach.
Identification of protein lysine methylation readers with a yeast three-hybrid approach.
Epigenetics & chromatin (2018-01-27)
Agnieszka Anna Rawłuszko-Wieczorek, Franziska Knodel, Raluca Tamas, Arunkumar Dhayalan, Albert Jeltsch
ABSTRACT
Protein posttranslational modifications (PTMs) occur broadly in the human proteome, and their biological outcome is often mediated indirectly by reader proteins that specifically bind to modified proteins and trigger downstream effects. Particularly, many lysine methylation sites among histone and nonhistone proteins have been characterized; however, the list of readers associated with them is incomplete. This study introduces a modified yeast three-hybrid system (Y3H) to screen for methyllysine readers. A lysine methyltransferase is expressed together with its target protein or protein domain functioning as bait, and a human cDNA library serves as prey. Proof of principle was established using H3K9me3 as a bait and known H3K9me3 readers like the chromodomains of CBX1 or MPP8 as prey. We next conducted an unbiased screen using a library composed of human-specific open reading frames. It led to the identification of already known lysine methylation-dependent readers and of novel methyllysine reader candidates, which were further confirmed by co-localization with H3K9me3 in human cell nuclei. Our approach introduces a cost-effective method for screening reading domains binding to histone and nonhistone proteins which is not limited by expression levels of the candidate reading proteins. Identification of already known and novel H3K9me3 readers proofs the power of the Y3H assay which will allow for proteome-wide screens of PTM readers.
Sigma-Aldrich
Glass beads, acid-washed, 425-600 μm (30-40 U.S. sieve)
Pricing and availability is not currently available.