Skip to Content
Merck

AMBH9884CA61

Ambeed, Inc.

4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

Sign In to View Organizational & Contract Pricing

Select a Size

Product AMBH9884CA61 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
282.53
MDL number:
Product AMBH9884CA61 is not currently sold in your country.

Iupac Name

4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

Inchi Code

1S/C13H16BClO4/c1-12(2)13(3,4)19-14(18-12)9-7-8(11(16)17)5-6-10(9)15/h5-7H,1-4H3,(H,16,17)

InChI key

MRHDRTNQFRBXFI-UHFFFAOYSA-N

storage temp.

Sealed in dry, room temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/918331-73-4.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/918331-73-4.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library