Skip to Content
Merck

Skip To

BDPH9BCDC8A0

Bide Pharmatech

(3-Methyl-4-(methylthio)phenyl)boronic acid

Synonym(s):

(3-methyl-4-(methylthio)phenyl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C8H11O2S1B1
Molecular Weight:
182.05
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H11BO2S/c1-6-5-7(9(10)11)3-4-8(6)12-2/h3-5,10-11H,1-2H3

Inchi Key

MKFDONWOZBIQGF-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library